Gene Transcripts
- Gene Symbol
- Rxfp1
- Entrez Gene ID
- 381489
- Gene Name
- relaxin/insulin-like family peptide receptor 1
- Gene Aliases
- Gm1018, Lgr7
- Chromosome Location
- Chr. 3: 79641600 - 79737951 on Build GRCm38
- UniGene ID
- Mm.331086
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK086423.1 |
- |
1 |
1947 |
|
AK142756.1 |
- |
1 |
1947 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BB525582.2 |
- |
1 |
472 |
|
Chr. 3 - 79641611 - 79642259 [-] |
View Full Product Details