Gene Transcripts
- Gene Symbol
- Gemin4
- Entrez Gene ID
- 276919
- Gene Name
- gem (nuclear organelle) associated protein 4
- Gene Aliases
- 4932415L08Rik, AU015359
- Chromosome Location
- Chr. 11: 76210571 - 76217572 on Build GRCm38
- UniGene ID
- Mm.482713
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK028076.1 |
- |
2 |
156 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BY706277.1 |
- |
2 |
156 |
|
Chr. 11 - 76217079 - 76217515 [-] |
View Full Product Details