Gene Transcripts
- Gene Symbol
- MUC19
- Entrez Gene ID
- 283463
- Gene Name
- mucin 19, oligomeric
- Gene Aliases
- MUC-19
- Chromosome Location
- Chr. 12: 40393394 - 40570757 on Build GRCh38
- UniGene ID
- Hs.632712
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
CR749517.1 |
- |
11 |
3221 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000474954.5 |
- |
11 |
3220 |
|
Chr. 12 - 40522485 - 40534803 [+] |
View Full Product Details