Gene Transcripts

Gene Symbol
SNAP25-AS1
Entrez Gene ID
100131208
Gene Name
SNAP25 antisense RNA 1
Gene Aliases
Chromosome Location
Chr. 20: 10023812 - 10219506 on Build GRCh38
UniGene ID
Hs.658178
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_040710.1 5 2690
GenBank mRNA AK124961.1 - 5 2687


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GENCODE ENST00000603542.5 - 5 2938 Chr. 20 - 10023812 - 10219754 [-]
GenBankmRNA CR609457.1 - 1 2571 Chr. 20 - 10023856 - 10026651 [-]

View Full Product Details