Gene Transcripts
- Gene Symbol
- TM4SF19
- Entrez Gene ID
- 116211
- Gene Name
- transmembrane 4 L six family member 19
- Gene Aliases
- OCTM4
- Chromosome Location
- Chr. 3: 196323546 - 196338420 on Build GRCh38
- UniGene ID
- Hs.733264
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
CA430273.1 |
- |
1 |
457 |
|
- Gene Symbol
- TM4SF19-AS1
- Entrez Gene ID
- 100874214
- Gene Name
- TM4SF19 antisense RNA 1
- Gene Aliases
- Chromosome Location
- Chr. 3: 196318332 - 196325570 on Build GRCh38
- UniGene ID
- Hs.672105
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_046724.1 |
|
2 |
261 |
|
GenBank mRNA |
AK124973.1 |
- |
2 |
243 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000452051.1 |
- |
2 |
243 |
|
Chr. 3 - 196318350 - 196325570 [+] |
GenBankmRNA |
DA376589.1 |
- |
2 |
243 |
|
Chr. 3 - 196318350 - 196324304 [+] |
View Full Product Details