Gene Transcripts
- Gene Symbol
- ANO1-AS2
- Entrez Gene ID
- 100009613
- Gene Name
- ANO1 antisense RNA 2 (head to head)
- Gene Aliases
- C11orf78, NCRNA00224
- Chromosome Location
- Chr. 11: 70072434 - 70075348 on Build GRCh38
- UniGene ID
- Hs.131238
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_103835.1 |
|
2 |
164 |
|
GenBank mRNA |
AW592641.1 |
- |
2 |
164 |
|
EF419887.1 |
- |
1 |
273 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000531347.1 |
- |
1 |
364 |
|
Chr. 11 - 70072434 - 70075433 [-] |
View Full Product Details