Gene Transcripts

Gene Symbol
Slc26a7
Entrez Gene ID
208890
Gene Name
solute carrier family 26, member 7
Gene Aliases
Chromosome Location
Chr. 4: 14502430 - 14621971 on Build GRCm38
UniGene ID
Mm.483839
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145947.2 NP_666059.2 2 250
GenBank mRNA AF345194.1 - 1 152
AK054327.1 - 2 251
BC026928.1 - 2 229 4020760

View Full Product Details