Gene Transcripts

Gene Symbol
Zmym5
Entrez Gene ID
219105
Gene Name
zinc finger, MYM-type 5
Gene Aliases
9830124H08Rik
Chromosome Location
Chr. 14: 56790585 - 56811896 on Build GRCm38
UniGene ID
Mm.473172
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001253752.1 NP_001240681.1 4 677
NM_001253753.1 NP_001240682.1 4 747
NM_144842.3 NP_659091.3 4 741
GenBank mRNA AK036513.1 - 3 603
AK155333.1 - 4 659
AK156501.1 - 4 749
BC027380.1 - 2 90
BC058644.1 - 3 538
BU518427.1 - 4 741

View Full Product Details