Gene Transcripts

Gene Symbol
Lrrc29
Entrez Gene ID
234684
Gene Name
leucine rich repeat containing 29
Gene Aliases
A630024J02Rik, FBL9, FBXL9
Chromosome Location
Chr. 8: 105312340 - 105326331 on Build GRCm38
UniGene ID
Mm.381717
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177449.3 NP_803232.2 5 1332
GenBank mRNA AK041606.1 - 6 1835
AK042435.1 - 6 1836
AK148120.1 - 5 1334
AK170400.1 - 3 3244

View Full Product Details