Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_004805.3 | NP_004796.1 | 2 | 271 | |
GenBank mRNA | BC017205.2 | - | 2 | 243 | |
BC093795.1 | - | 2 | 271 | 7939640 | |
BC093797.1 | - | 2 | 273 | 7939642 | |
U85510.1 | - | 2 | 239 |