Gene Transcripts

Gene Symbol
Slc22a15
Entrez Gene ID
242126
Gene Name
solute carrier family 22 (organic anion/cation transporter), member 15
Gene Aliases
2610034P21Rik, 4930477M19, A530052I06Rik, BB219042
Chromosome Location
Chr. 3: 101855768 - 101924453 on Build GRCm38
UniGene ID
Mm.90204
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039371.2 NP_001034460.2 7 1405
GenBank mRNA AK029695.1 - 4 563
BC094925.1 - 7 1294
BC152347.1 - 2 300

View Full Product Details