Gene Transcripts

Gene Symbol
Mfsd7a
Entrez Gene ID
243197
Gene Name
major facilitator superfamily domain containing 7A
Gene Aliases
4732482E20Rik, Mfsd7
Chromosome Location
Chr. 5: 108441052 - 108450260 on Build GRCm38
UniGene ID
Mm.106818
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172883.2 NP_766471.1 2 193
GenBank mRNA AK029021.1 - 2 193
AK087226.1 - 2 403
AK171069.1 - 1 2825

View Full Product Details