Gene Transcripts

Gene Symbol
Vmn1r71
Entrez Gene ID
252910
Gene Name
vomeronasal 1 receptor 71
Gene Aliases
V1re13
Chromosome Location
Chr. 7: 10747502 - 10749538 on Build GRCm38
UniGene ID
Mm.451639
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145848.3 NP_665847.1 1 1137
GenBank mRNA AF394948.1 - 1 1135

View Full Product Details