Gene Transcripts

Gene Symbol
Ttc26
Entrez Gene ID
264134
Gene Name
tetratricopeptide repeat domain 26
Gene Aliases
9330141E21Rik, 9430097H08Rik
Chromosome Location
Chr. 6: 38381474 - 38427647 on Build GRCm38
UniGene ID
Mm.253446
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153600.2 NP_705828.2 6 561
GenBank mRNA AK035185.1 - 6 569
BC066060.1 - 6 561 6848602
BC066087.1 - 7 751

View Full Product Details