Gene Transcripts

Gene Symbol
Serpina5
Entrez Gene ID
268591
Gene Name
serine (or cysteine) peptidase inhibitor, clade A, member 5
Gene Aliases
4933415L04, PAI-3, Pci
Chromosome Location
Chr. 12: 104099313 - 104106138 on Build GRCm38
UniGene ID
Mm.73172
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172953.3 NP_766541.2 2 749
GenBank mRNA AK077160.1 - 2 750
AK087714.1 - 2 712
BC090981.1 - 2 712 30285598

View Full Product Details