Gene Transcripts

Gene Symbol
Lrrc16b
Entrez Gene ID
268747
Gene Name
leucine rich repeat containing 16B
Gene Aliases
mFLJ00240
Chromosome Location
Chr. 14: 55490656 - 55508272 on Build GRCm38
UniGene ID
Mm.151577
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001024645.1 NP_001019816.1 39 4208
GenBank mRNA AK131164.1 - 38 4254
AK158466.1 - 39 4690
AK161800.1 - 2 1682
BC036313.1 - 21 3440
BC060252.1 - 20 2748
BC082805.1 - 38 4369
BC092223.1 - 37 4465

View Full Product Details