Gene Transcripts

Gene Symbol
Treml2
Entrez Gene ID
328833
Gene Name
triggering receptor expressed on myeloid cells-like 2
Gene Aliases
AW049306, Gm750, Tlt2
Chromosome Location
Chr. 17: 48299387 - 48312534 on Build GRCm38
UniGene ID
Mm.329104
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033405.2 NP_001028577.2 3 997
GenBank mRNA DQ087184.1 - 3 521
DQ341272.1 - 3 1096

View Full Product Details