Gene Transcripts

Gene Symbol
Noto
Entrez Gene ID
384452
Gene Name
notochord homolog (Xenopus laevis)
Gene Aliases
Flh, MmNot, Not, tc
Chromosome Location
Chr. 6: 85423886 - 85428877 on Build GRCm38
UniGene ID
Mm.347883
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001007472.2 NP_001007473.1 2, 3 690
GenBank mRNA AJ862873.1 - 2, 3 647
AJ862874.1 - 2, 3 647
AJ862875.1 - 2, 3 647
AK135886.1 - 2, 3 690
BC139154.1 - 2, 3 634 8862176
BC139155.1 - 2, 3 634 8862177

View Full Product Details