Gene Transcripts

Gene Symbol
Ugt2a2
Entrez Gene ID
552899
Gene Name
UDP glucuronosyltransferase 2 family, polypeptide A2
Gene Aliases
Chromosome Location
Chr. 5: 87459493 - 87482258 on Build GRCm38
UniGene ID
Mm.26794
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001024148.1 NP_001019319.1 2 680
GenBank mRNA BC048920.1 - 1 681
BC058786.1 - 2 680 6310060

View Full Product Details