Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_003039.2 | NP_003030.1 | 6 | 953 | |
XM_005263491.3 | XP_005263548.1 | 7 | 751 | ||
XM_017002133.1 | XP_016857622.1 | 8 | 979 | ||
XM_017002134.1 | XP_016857623.1 | 8 | 945 | ||
XM_017002135.1 | XP_016857624.1 | 8 | 1039 | ||
XM_017002136.1 | XP_016857625.1 | 8 | 1015 | ||
XM_017002137.1 | XP_016857626.1 | 9 | 1051 | ||
XM_017002138.1 | XP_016857627.1 | 9 | 1027 | ||
XM_017002139.1 | XP_016857628.1 | 7 | 961 | ||
XM_017002140.1 | XP_016857629.1 | 8 | 882 | ||
XM_017002141.1 | XP_016857630.1 | 7 | 784 | ||
XM_017002142.1 | XP_016857631.1 | 4 | 524 | ||
GenBank mRNA | AK290398.1 | - | 6 | 743 | |
AK294368.1 | - | 6 | 739 | ||
AK300091.1 | - | 4 | 410 | ||
AK300477.1 | - | 4 | 529 | ||
AK302047.1 | - | 6 | 594 | ||
AK302057.1 | - | 9 | 877 | ||
AK302137.1 | - | 8 | 829 | ||
BC001692.2 | - | 6 | 712 | 2966459 | |
BC001820.2 | - | 6 | 712 | 2966459 | |
BC035878.1 | - | 5 | 2335 | ||
M55531.1 | - | 6 | 736 |