Gene Transcripts
- Gene Symbol
- TMSB4X
- Entrez Gene ID
- 7114
- Gene Name
- thymosin beta 4, X-linked
- Gene Aliases
- FX, PTMB4, TB4X, TMSB4
- Chromosome Location
- Chr. X: 12975107 - 12977227 on Build GRCh38
- UniGene ID
- Hs.437277
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_021109.3 |
NP_066932.1
|
1 |
173 |
|
GenBank mRNA |
AK055976.1 |
- |
1, 2 |
1247 |
|
AK057827.1 |
- |
2, 3 |
170 |
|
AK311747.1 |
- |
2, 3 |
171 |
|
AV715954.1 |
- |
2, 3 |
173 |
|
BC001631.1 |
- |
1 |
154 |
|
BC016732.1 |
- |
1 |
174 |
|
BC022857.1 |
- |
1 |
174 |
|
BC061586.1 |
- |
2, 3 |
160 |
|
BC062436.1 |
- |
1 |
174 |
|
BC064954.1 |
- |
2, 3 |
154 |
|
BC070248.1 |
- |
2, 3 |
171 |
|
BC070249.1 |
- |
1 |
170 |
|
BC083509.1 |
- |
1 |
170 |
|
BC092437.1 |
- |
1 |
170 |
|
BC101790.1 |
- |
2, 3 |
157 |
8069296 |
BC101792.1 |
- |
2, 3 |
157 |
8069298 |
BC104464.1 |
- |
1 |
124 |
40031330 |
BC104465.1 |
- |
1 |
124 |
40031337 |
BC104666.1 |
- |
2, 3 |
174 |
|
BC107789.1 |
- |
2, 3 |
177 |
|
BC128102.1 |
- |
2, 3 |
142 |
40112039 |
BC128103.1 |
- |
2, 3 |
145 |
40112043 |
BC139925.1 |
- |
1 |
159 |
40148985 |
BC141975.1 |
- |
2, 3 |
145 |
40112044 |
BC141976.1 |
- |
2, 3 |
145 |
40112045 |
BC151215.1 |
- |
1 |
160 |
40148015 |
BT007090.1 |
- |
1, 2 |
95 |
|
M17733.1 |
- |
2, 3 |
172 |
|
X02493.1 |
- |
1, 2 |
75 |
|
- Gene Symbol
- TMSB4XP1
- Entrez Gene ID
- 7115
- Gene Name
- thymosin beta 4, X-linked pseudogene 1
- Gene Aliases
- TMSL1
- Chromosome Location
- Chr. 1: 42499781 - 42500417 on Build GRCh38
- UniGene ID
- Hs.655548
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC090885.1 |
- |
1 |
34 |
|
View Full Product Details