Gene Transcripts

Gene Symbol
Pard3b
Entrez Gene ID
301455
Gene Name
par-3 family cell polarity regulator beta
Gene Aliases
Chromosome Location
Chr. 9: 68414339 - 69426594 on Build Rnor_6.0
UniGene ID
Rn.214104
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001191808.1 NP_001178737.1 8 1068
XM_008767088.1 XP_008765310.1 7 894

View Full Product Details