Gene Transcripts

Gene Symbol
Scarf1
Entrez Gene ID
303313
Gene Name
scavenger receptor class F, member 1
Gene Aliases
Chromosome Location
Chr. 10: 63662967 - 63683916 on Build Rnor_6.0
UniGene ID
Rn.224782
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107022.1 NP_001100492.1 262
GenBank mRNA BC088259.1 - 3 248
FQ234496.1 - 3 278

View Full Product Details