Gene Transcripts

Gene Symbol
Lrfn5
Entrez Gene ID
314164
Gene Name
leucine rich repeat and fibronectin type III domain containing 5
Gene Aliases
Chromosome Location
Chr. 6: 83083740 - 83438602 on Build Rnor_6.0
UniGene ID
Rn.224998
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108024.1 NP_001101494.1 3 953
XM_008764687.1 XP_008762909.1 1 553

View Full Product Details