Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_003745.1 | NP_003736.1 | 2 | 1088 | |
GenBank mRNA | AB000676.1 | - | 1 | 892 | |
AB000734.1 | - | 2 | 1088 | ||
AB005043.1 | - | 1 | 987 | ||
AK127621.1 | - | 1 | 1656 | ||
U88326.1 | - | 1 | 957 |