Gene Transcripts

Gene Symbol
Sall3
Entrez Gene ID
364910
Gene Name
spalt-like transcription factor 3
Gene Aliases
Chromosome Location
Chr. 18: 77572200 - 77591685 on Build Rnor_6.0
UniGene ID
Rn.99341
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108892.1 NP_001102362.1 1 968
XM_006254985.2 XP_006255047.1 2 1083
XM_006254987.2 XP_006255049.1 2 1083

View Full Product Details