Gene Transcripts

Gene Symbol
Mb21d2
Entrez Gene ID
498100
Gene Name
Mab-21 domain containing 2
Gene Aliases
RGD1559643
Chromosome Location
Chr. 11: 75434198 - 75533476 on Build Rnor_6.0
UniGene ID
Rn.11815
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109056.2 NP_001102526.2 2 1449
XM_006248523.2 XP_006248585.1 2 2972
XM_006248524.2 XP_006248586.1 2 1222

View Full Product Details