Gene Transcripts

Gene Symbol
Spesp1
Entrez Gene ID
501010
Gene Name
sperm equatorial segment protein 1
Gene Aliases
RGD1563468
Chromosome Location
Chr. 8: 67217801 - 67233176 on Build Rnor_6.0
UniGene ID
Rn.123383
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001077682.1 NP_001071150.1 2 812
XM_006243210.2 XP_006243272.1 2 876
GenBank mRNA BC127527.1 - 2 812

View Full Product Details