Gene Transcripts
- Gene Symbol
- Ndufa10l1
- Entrez Gene ID
- 316632
- Gene Name
- NADH dehydrogenase (ubiquinone) 1 alpha subcomplex 10-like 1
- Gene Aliases
- Ndufa10
- Chromosome Location
- Chr. 6: 62796951 - 62798384 on Build Rnor_6.0
- UniGene ID
- Rn.102304
- Species
- Rattus norvegicus
- Gene Symbol
- Ndufa10
- Entrez Gene ID
- 678759
- Gene Name
- NADH dehydrogenase (ubiquinone) 1 alpha subcomplex 10
- Gene Aliases
- ndufa10l
- Chromosome Location
- Chr. 9: 99617051 - 99651825 on Build Rnor_6.0
- UniGene ID
- Rn.169414
- Species
- Rattus norvegicus
View Full Product Details