Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_004863.3 | NP_004854.1 | 5 | 903 | |
XM_011537384.2 | XP_011535686.1 | 5 | 799 | ||
GenBank mRNA | AB011098.1 | - | 5 | 903 | |
BC005123.2 | - | 5 | 804 | 3688136 | |
BQ338973.1 | - | 2 | 235 | ||
U15555.1 | - | 4 | 539 | ||
Y08686.1 | - | 5 | 763 |