Gene Transcripts

Gene Symbol
LOC690319
Entrez Gene ID
690319
Gene Name
similar to Alpha-1-antitrypsin-related protein precursor
Gene Aliases
Chromosome Location
Chr. 6: 127590734 - 127600821 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_001074075.4 XP_001074075.2 2 573

View Full Product Details