Gene Transcripts
- Gene Symbol
- OR2T3
- Entrez Gene ID
- 343173
- Gene Name
- olfactory receptor family 2 subfamily T member 3
- Gene Aliases
- Chromosome Location
- Chr. 1: 248473351 - 248474307 on Build GRCh38
- UniGene ID
- Hs.554481
- Species
- Homo sapiens
- Gene Symbol
- OR2T34
- Entrez Gene ID
- 127068
- Gene Name
- olfactory receptor family 2 subfamily T member 34
- Gene Aliases
- Chromosome Location
- Chr. 1: 248573801 - 248574757 on Build GRCh38
- UniGene ID
- Hs.553584
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC136924.1 |
- |
1 |
828 |
|
BC136926.1 |
- |
1 |
828 |
9020918 |
View Full Product Details