Gene Transcripts

Gene Symbol
Speer3
Entrez Gene ID
71026
Gene Name
spermatogenesis associated glutamate (E)-rich protein 3
Gene Aliases
4933405P08Rik, SPEER-3
Chromosome Location
Chr. 5: 13791619 - 13796819 on Build GRCm38
UniGene ID
Mm.158536
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027650.3 NP_081926.2 2 229
GenBank mRNA AK016676.1 - 2 216
CJ042389.1 - 2 229

View Full Product Details