Gene Transcripts

Gene Symbol
BTN3A1
Entrez Gene ID
11119
Gene Name
butyrophilin subfamily 3 member A1
Gene Aliases
BT3.1, BTF5, BTN3.1, CD277
Chromosome Location
Chr. 6: 26402237 - 26415216 on Build GRCh38
UniGene ID
Hs.191510
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001145008.1 NP_001138480.1 10 1646
NM_001145009.1 NP_001138481.1 10 2559
NM_007048.5 NP_008979.3 10 1802
NM_194441.2 NP_919423.1 10 2096
XM_005248833.2 XP_005248890.1 9 1749
XM_005248834.3 XP_005248891.1 10 2261
XM_006714983.2 XP_006715046.1 9 1507
GenBank mRNA AK223474.1 - 9 1583
AK290193.1 - 10 1799
AK298652.1 - 10 1531
AK299327.1 - 10 1588
BC118586.1 - 10 1763 40085016
BC121800.1 - 10 1763 40085015
CB148209.1 - 6 537
DA044176.1 - 1 550
U90552.2 - 10 1732

View Full Product Details