Gene Transcripts

Gene Symbol
Dcaf4
Entrez Gene ID
362762
Gene Name
DDB1 and CUL4 associated factor 4
Gene Aliases
Wdr21
Chromosome Location
Chr. 6: 107001763 - 107027466 on Build Rnor_6.0
UniGene ID
Rn.24961
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108716.1 NP_001102186.1 5 521
XM_006240347.2 XP_006240409.1 5 521
XM_008764814.1 XP_008763036.1 4 569

View Full Product Details