Gene Transcripts

Gene Symbol
TMSB15B
Entrez Gene ID
286527
Gene Name
thymosin beta 15B
Gene Aliases
Tbeta15b
Chromosome Location
Chr. X: 103962627 - 103965990 on Build GRCh38
UniGene ID
Hs.56145
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_194324.2 NP_919305.2 3 381
GenBank mRNA AK291479.1 - 3 357
BC093093.1 - 3 381

View Full Product Details