Gene Transcripts

Gene Symbol
Cdk16
Entrez Gene ID
18555
Gene Name
cyclin-dependent kinase 16
Gene Aliases
Crk5, Pctaire1, Pctk1
Chromosome Location
Chr. X: 20687959 - 20699880 on Build GRCm38
UniGene ID
Mm.102574
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011049.4 NP_035179.1 7 1032
GenBank mRNA AK051425.1 - 7 1033
AK141083.1 - 1 51
AK143518.1 - 7 1060
AK145483.1 - 7 1062
AK166188.1 - 7 1241
AK179102.1 - 1 85
AK180401.1 - 1 85
AK180999.1 - 1 85
AK194254.1 - 1 85
AK194908.1 - 1 85
AK203854.1 - 1 85
AK214881.1 - 2 148
BC011069.1 - 7 775 4006745
BC013663.1 - 7 1091 4208974
X69025.1 - 7 1054

View Full Product Details