Gene Transcripts

Gene Symbol
CHST5
Entrez Gene ID
23563
Gene Name
carbohydrate sulfotransferase 5
Gene Aliases
I-GlcNAc-6-ST, I-GlcNAc6ST, glcNAc6ST-3, gn6st-3, hIGn6ST
Chromosome Location
Chr. 16: 75528530 - 75535170 on Build GRCh38
UniGene ID
Hs.745424
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024533.4 NP_078809.2 3 1904
GenBank mRNA AF246718.1 - 3 1905
BC147002.1 - 1 729 9056930
BC147007.1 - 1 729 9056935

View Full Product Details