Gene Transcripts
- Gene Symbol
- SNX11
- Entrez Gene ID
- 29916
- Gene Name
- sorting nexin 11
- Gene Aliases
- Chromosome Location
- Chr. 17: 48107533 - 48123064 on Build GRCh38
- UniGene ID
- Hs.15827
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_013323.2 |
NP_037455.2
|
3, 4 |
391 |
|
NM_152244.1 |
NP_689450.1
|
4, 5 |
478 |
|
XM_005257260.3 |
XP_005257317.1
|
3, 4 |
158 |
|
XM_005257261.3 |
XP_005257318.1
|
3, 4 |
425 |
|
XM_005257262.3 |
XP_005257319.1
|
3, 4 |
219 |
|
XM_011524697.2 |
XP_011522999.1
|
4, 5 |
261 |
|
GenBank mRNA |
AF121861.1 |
- |
3, 4 |
411 |
|
AK023932.1 |
- |
4, 5 |
478 |
|
AK091852.1 |
- |
4, 5 |
376 |
|
AK298551.1 |
- |
3, 4 |
214 |
|
AK316374.1 |
- |
3, 4 |
211 |
|
BC000768.2 |
- |
3, 4 |
208 |
2963905 |
BC103721.1 |
- |
3, 4 |
197 |
6162638 |
BT006723.1 |
- |
2, 3 |
124 |
|
View Full Product Details