Gene Transcripts
- Gene Symbol
- MBTD1
- Entrez Gene ID
- 54799
- Gene Name
- mbt domain containing 1
- Gene Aliases
- SA49P01
- Chromosome Location
- Chr. 17: 51177419 - 51260511 on Build GRCh38
- UniGene ID
- Hs.656803
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_017643.2 |
NP_060113.2
|
14, 15 |
1807 |
|
XM_005257467.4 |
XP_005257524.1
|
13, 14 |
1752 |
|
XM_005257468.4 |
XP_005257525.1
|
13, 14 |
1745 |
|
XM_011524922.2 |
XP_011523224.1
|
13, 14 |
1754 |
|
XM_011524923.2 |
XP_011523225.1
|
13, 14 |
1751 |
|
XM_011524924.2 |
XP_011523226.1
|
13, 14 |
1747 |
|
XM_011524925.1 |
XP_011523227.1
|
14, 15 |
1868 |
|
XM_011524926.2 |
XP_011523228.1
|
14, 15 |
1972 |
|
XM_011524928.2 |
XP_011523230.1
|
13, 14 |
1909 |
|
XM_011524929.2 |
XP_011523231.1
|
14, 15 |
1823 |
|
XM_011524930.1 |
XP_011523232.1
|
13, 14 |
1545 |
|
XM_011524931.2 |
XP_011523233.1
|
12, 13 |
1379 |
|
XM_011524933.2 |
XP_011523235.1
|
15, 16 |
1929 |
|
XM_011524934.2 |
XP_011523236.1
|
12, 13 |
1605 |
|
XM_011524935.2 |
XP_011523237.1
|
12, 13 |
1598 |
|
XM_011524936.1 |
XP_011523238.1
|
12, 13 |
1397 |
|
XM_017024782.1 |
XP_016880271.1
|
14, 15 |
2119 |
|
GenBank mRNA |
AK000062.1 |
- |
10, 11 |
1139 |
|
AK124061.1 |
- |
12, 13 |
1397 |
|
BC034364.1 |
- |
14, 15 |
1778 |
|
BC101736.1 |
- |
9, 10 |
1068 |
8069242 |
View Full Product Details