Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_018228.2 | NP_060698.2 | 2 | 1488 | |
XM_011536911.1 | XP_011535213.1 | 3 | 1535 | ||
GenBank mRNA | AK001673.1 | - | 2 | 1442 | |
AK023213.1 | - | 2 | 1455 | ||
BC053325.1 | - | 2 | 1471 | 6577658 |