Gene Transcripts

Gene Symbol
MRGBP
Entrez Gene ID
55257
Gene Name
MRG/MORF4L binding protein
Gene Aliases
C20orf20, Eaf7, MRG15BP, URCC4
Chromosome Location
Chr. 20: 62796405 - 62801738 on Build GRCh38
UniGene ID
Hs.443260
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018270.5 NP_060740.1 5 584
NR_136405.1 5 706
GenBank mRNA AB085682.1 - 5 484
AK001776.1 - 5 536
BC009889.2 - 5 503 4026378
BC014235.2 - 5 508 4555060
BC018841.2 - 5 503 3050866
BM920599.1 - 612
BU902487.1 - 5 567

View Full Product Details