Gene Transcripts

Gene Symbol
LMO3
Entrez Gene ID
55885
Gene Name
LIM domain only 3
Gene Aliases
RBTN3, RBTNL2, RHOM3, Rhom-3
Chromosome Location
Chr. 12: 16548372 - 16610233 on Build GRCh38
UniGene ID
Hs.504908
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001001395.2 NP_001001395.1 2 447
NM_001243609.1 NP_001230538.1 3 514
NM_001243610.1 NP_001230539.1 3 286
NM_001243611.1 NP_001230540.1 2 683
NM_001243612.1 NP_001230541.1 2 371
NM_018640.4 NP_061110.2 2 705
NR_045012.1 3 514
XM_006719110.3 XP_006719173.1 3 522
XM_006719111.2 XP_006719174.1 2 565
XM_011520762.1 XP_011519064.1 3 413
XM_011520763.1 XP_011519065.1 3 320
XM_017019643.1 XP_016875132.1 2 668
GenBank mRNA AB044745.1 - 2 451
AB044746.1 - 4 352
AK095595.1 - 2 323
AK294474.1 - 2 371
AK294651.1 - 2 360
AK308594.1 - 3 425
AK308826.1 - 4 390
AK308896.1 - 3 326
AK312418.1 - 2 589
AK316021.1 - 2 307
BC026311.1 - 2 601 4799836
BC050085.1 - 3 287 5274811
BI553855.1 - 2 344
DA780743.1 - 2 448

View Full Product Details