Gene Transcripts

Gene Symbol
MIER1
Entrez Gene ID
57708
Gene Name
MIER1 transcriptional regulator
Gene Aliases
ER1, MI-ER1
Chromosome Location
Chr. 1: 66924895 - 66988619 on Build GRCh38
UniGene ID
Hs.605432
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001077700.2 NP_001071168.2 12, 13 1372
NM_001077701.2 NP_001071169.1 11, 12 1228
NM_001077702.2 NP_001071170.2 13, 14 1446
NM_001077703.2 NP_001071171.2 12, 13 1372
NM_001077704.2 NP_001071172.1 11, 12 1228
NM_001146110.1 NP_001139582.1 13, 14 1449
NM_001146111.1 NP_001139583.1 13, 14 1446
NM_001146112.1 NP_001139584.1 13, 14 1407
NM_001146113.1 NP_001139585.1 11, 12 1226
NM_020948.3 NP_065999.2 13, 14 1446
XM_005271076.3 XP_005271133.1 13, 14 1484
XM_011541864.2 XP_011540166.1 14, 15 1391
XM_011541865.2 XP_011540167.1 14, 15 1352
XM_017001919.1 XP_016857408.1 14, 15 1368
XM_017001920.1 XP_016857409.1 14, 15 1385
XM_017001921.1 XP_016857410.1 14, 15 1413
XM_017001922.1 XP_016857411.1 14, 15 1388
XM_017001923.1 XP_016857412.1 12, 13 1208
XM_017001924.1 XP_016857413.1 13, 14 1340
XM_017001925.1 XP_016857414.1 13, 14 1445
XM_017001926.1 XP_016857415.1 14, 15 1384
XM_017001927.1 XP_016857416.1 12, 13 1257
XM_017001928.1 XP_016857417.1 12, 13 1333
XM_017001929.1 XP_016857418.1 13, 14 1314
XM_017001930.1 XP_016857419.1 12, 13 1258
XM_017001931.1 XP_016857420.1 13, 14 1292
XM_017001932.1 XP_016857421.1 11, 12 1221
XM_017001933.1 XP_016857422.1 10, 11 1075
XM_017001934.1 XP_016857423.1 9, 10 971
GenBank mRNA AB046830.1 - 9, 10 997
AF515446.1 - 11, 12 1228
AF515447.1 - 12, 13 1309
AF515448.2 - 13, 14 1383
AK302061.1 - 12, 13 1372
AK307717.1 - 13, 14 1343
AL831987.2 - 13, 14 1388
AY124186.2 - 13, 14 1383
AY124187.1 - 12, 13 1309
AY124188.1 - 11, 12 1228
AY124189.2 - 13, 14 1383
AY124190.1 - 12, 13 1309
AY124191.1 - 11, 12 1228
AY124192.2 - 13, 14 1383
AY124193.1 - 12, 13 1309
AY124194.1 - 11, 12 1228
BC108726.1 - 13, 14 1327 5194296
BC125217.1 - 10, 11 1024 40123797
BC125218.1 - 10, 11 1024 40123803
CR627441.1 - 13, 14 1283

View Full Product Details