Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001018039.1 | NP_001018049.1 | 7, 8 | 969 | |
NM_001029880.2 | NP_001025051.1 | 7, 8 | 1071 | ||
XM_005252551.2 | XP_005252608.1 | 7, 8 | 852 | ||
XM_006717490.2 | XP_006717553.1 | 5, 6 | 844 | ||
XM_011519611.1 | XP_011517913.1 | 7, 8 | 1070 | ||
XM_017016466.1 | XP_016871955.1 | 7, 8 | 972 | ||
GenBank mRNA | AB046837.1 | - | 7, 8 | 969 | |
BC144327.1 | - | 7, 8 | 1036 | ||
BC152430.1 | - | 7, 8 | 969 | 8862546 |