Gene Transcripts

Gene Symbol
RXFP1
Entrez Gene ID
59350
Gene Name
relaxin/insulin like family peptide receptor 1
Gene Aliases
LGR7, RXFPR1
Chromosome Location
Chr. 4: 158521711 - 158653372 on Build GRCh38
UniGene ID
Hs.591686
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001253727.1 NP_001240656.1 12 1259
NM_001253728.1 NP_001240657.1 11 1079
NM_001253730.1 NP_001240659.1 11 1103
NM_001253732.1 NP_001240661.1 13 1262
NM_001253733.1 NP_001240662.1 12 1190
NM_021634.3 NP_067647.2 12 1178
NR_045579.1 12 1795
NR_045580.1 12 1231
NR_045581.1 12 1202
NR_045583.1 11 1121
NR_045584.1 13 1231
XM_011532174.1 XP_011530476.1 12 1228
XM_011532175.1 XP_011530477.1 11 1156
XM_011532176.1 XP_011530478.1 11 1078
XM_011532179.2 XP_011530481.1 12 1228
XM_017008517.1 XP_016864006.1 11 1156
XM_017008518.1 XP_016864007.1 12 1150
XM_017008519.1 XP_016864008.1 13 1192
XM_017008520.1 XP_016864009.1 12 1281
XM_017008521.1 XP_016864010.1 13 1111
XM_017008522.1 XP_016864011.1 12 1199
XM_017008523.1 XP_016864012.1 12 1150
XM_017008524.1 XP_016864013.1 11 1078
XM_017008525.1 XP_016864014.1 11 1082
XM_017008526.1 XP_016864015.1 7 649
GenBank mRNA AB208943.1 - 13 1000
AF190500.1 - 12 915
AK122647.1 - 14 1219
AK294694.1 - 12 1023
AK295040.1 - 12 1618
AK300379.1 - 11 974
AK307889.1 - 11 1020
AK312470.1 - 12 1022
AY899848.1 - 13 968
AY899850.1 - 11 816
BC112142.1 - 12 997 8327610
BC113617.1 - 12 1000 8322669
BC143556.1 - 12 1020
BC143558.1 - 12 1053
BC143559.1 - 11 925
BX647975.1 - 12 1116
BX647980.1 - 12 1116
BX647985.1 - 12 1145

View Full Product Details