Gene Transcripts
- Gene Symbol
- RXFP1
- Entrez Gene ID
- 59350
- Gene Name
- relaxin/insulin like family peptide receptor 1
- Gene Aliases
- LGR7, RXFPR1
- Chromosome Location
- Chr. 4: 158521711 - 158653372 on Build GRCh38
- UniGene ID
- Hs.591686
- Species
- Homo sapiens
View Full Product Details