Gene Transcripts

Gene Symbol
MS4A6A
Entrez Gene ID
64231
Gene Name
membrane spanning 4-domains A6A
Gene Aliases
4SPAN3, 4SPAN3.2, CD20L3, CDA01, MS4A6, MST090, MSTP090
Chromosome Location
Chr. 11: 60171607 - 60184666 on Build GRCh38
UniGene ID
Hs.523702
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001247999.1 NP_001234928.1 4 768
NM_022349.3 NP_071744.2 3 579
NM_152851.2 NP_690590.1 3 579
NM_152852.2 NP_690591.1 4 704
XM_005274177.2 XP_005274234.1 4 763
XM_006718658.2 XP_006718721.1 4 638
XM_011545209.1 XP_011543511.1 4 638
XM_017018125.1 XP_016873614.1 4 638
GenBank mRNA AB013104.1 - 3 287
AF142409.1 - 3 307
AF172819.1 - 4 725
AF212240.1 - 3 448
AF237908.1 - 3 449
AF253977.1 - 3 449
AF286866.1 - 2 227
AF350502.1 - 3 296
AF350503.1 - 3 296
AF354930.1 - 3 449
AK026454.1 - 2 2017
AK291870.1 - 4 704
AK300725.1 - 3 313
BC022854.1 - 3 318 3934920
BC108728.1 - 3 352 6196213
DA917243.1 - 3 403
DA979138.1 - 3 350

View Full Product Details