Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001134774.1 | NP_001128246.1 | 8 | 1136 | |
NM_001134775.1 | NP_001128247.1 | 9 | 1430 | ||
NM_001134776.1 | NP_001128248.1 | 9 | 1309 | ||
NM_001318734.1 | NP_001305663.1 | 9 | 1367 | ||
NM_022822.2 | NP_073733.1 | 9 | 1404 | ||
XM_005274183.1 | XP_005274240.1 | 9 | 1346 | ||
XM_005274184.1 | XP_005274241.1 | 9 | 1367 | ||
XM_017018133.1 | XP_016873622.1 | 7 | 1303 | ||
GenBank mRNA | AK022449.1 | - | 9 | 1296 | |
AK022907.1 | - | 9 | 1349 | ||
AK094593.1 | - | 7 | 1006 | ||
AK126489.1 | - | 7 | 1788 | ||
AK315725.1 | - | 9 | 1299 | ||
AL136864.1 | - | 9 | 1330 | ||
BC034373.1 | - | 9 | 1292 | 4342879 |