Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_025235.3 | NP_079511.1 | 6 | 973 | |
XM_005270185.4 | XP_005270242.2 | 6 | 1004 | ||
XM_011540213.1 | XP_011538515.1 | 6 | 1004 | ||
XM_017016696.1 | XP_016872185.1 | 6 | 941 | ||
XM_017016697.1 | XP_016872186.1 | 6 | 957 | ||
XM_017016698.1 | XP_016872187.1 | 6 | 668 | ||
XM_017016699.1 | XP_016872188.1 | 6 | 957 | ||
XM_017016700.1 | XP_016872189.1 | 6 | 554 | ||
XM_017016701.1 | XP_016872190.1 | 6 | 1004 | ||
GenBank mRNA | AF264912.1 | - | 6 | 676 | |
AF305081.1 | - | 6 | 978 | ||
AF309033.1 | - | 6 | 927 | ||
AF329696.1 | - | 6 | 815 | ||
AF342982.1 | - | 6 | 676 | ||
AF438201.1 | - | 6 | 1055 | ||
AK314612.1 | - | 6 | 941 |